Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lea-1
Entrez Gene ID
3564838
Gene Name
protein-coding
Gene Aliases
CELE_K08H10.1
Chromosome Location
Chr.V: 10011923 - 10028168 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269232.3 NP_001256161.1 8 - 9 3456 79
NM_001269235.3 NP_001256164.1 7 - 8 3408 79
NM_001269241.4 NP_001256170.1 6 - 7 3192 79
NM_001269245.3 NP_001256174.1 2 - 3 1305 79
NM_001373085.2 NP_001360513.1 9 - 9 3828 79
NM_001373087.2 NP_001360515.1 8 - 8 3780 79
NM_001373089.2 NP_001360517.1 7 - 7 3564 79

View Full Product Details