Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
agr-1
Entrez Gene ID
3565243
Gene Name
AGRin (synaptic protein) homolog
Gene Aliases
CELE_F41G3.12
Chromosome Location
Chr.II: 6760674 - 6775176 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026981.6 NP_001022152.3 1 - 2 400 153
GenBank mRNA AM773423.1 - 1 - 2 400 153

View Full Product Details