Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mig-14
Entrez Gene ID
175020
Gene Name
Protein wntless homolog
Gene Aliases
CELE_R06B9.6
Chromosome Location
Chr.II: 13741738 - 13749983 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027104.4 NP_001022275.1 8 - 9 1552 58

View Full Product Details