Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
aqp-12
Entrez Gene ID
3565789
Gene Name
AQuaPorin or aquaglyceroporin related
Gene Aliases
CELE_Y57A10A.35
Chromosome Location
Chr.II: 12203221 - 12209166 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027309.3 NP_001022480.1 1 - 2 204 65

View Full Product Details