Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
zfp-1
Entrez Gene ID
176237
Gene Name
protein-coding
Gene Aliases
CELE_F54F2.2
Chromosome Location
Chr.III: 8791545 - 8806289 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027439.2 NP_001022610.1 5 - 6 1566 93

View Full Product Details