Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lgc-43
Entrez Gene ID
259591
Gene Name
Neur_chan_LBD domain-containing protein
Gene Aliases
CELE_C43F9.9
Chromosome Location
Chr.IV: 10578667 - 10581801 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027891.3 NP_001023062.1 5 - 6 1058 118

View Full Product Details