Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fkb-2
Entrez Gene ID
173160
Gene Name
Peptidylprolyl isomerase
Gene Aliases
CELE_Y18D10A.19
Chromosome Location
Chr.I: 12918722 - 12919287 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026551.6 NP_001021722.1 1 - 2 201 102

View Full Product Details