Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-72
Entrez Gene ID
191721
Gene Name
Nuclear Hormone Receptor family
Gene Aliases
CELE_C17A2.8
Chromosome Location
Chr.II: 3849047 - 3850755 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062270.6 NP_494671.2 3 - 4 432 147

View Full Product Details