Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
soc-2
Entrez Gene ID
177286
Gene Name
protein-coding
Gene Aliases
CELE_AC7.2
Chromosome Location
Chr.IV: 5127006 - 5152491 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306829.3 NP_001293758.1 1 - 1 195 70

View Full Product Details