Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aqp-4
Entrez Gene ID
179366
Gene Name
AQuaPorin or aquaglyceroporin related
Gene Aliases
CELE_F40F9.9
Chromosome Location
Chr.V: 9734938 - 9739740 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073111.7 NP_505512.3 3 - 4 558 100

View Full Product Details