Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srw-31
Entrez Gene ID
186943
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_K03D7.2
Chromosome Location
Chr.V: 17506589 - 17508506 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075034.2 NP_507435.2 3 - 4 933 126

View Full Product Details