Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rpl-2
Entrez Gene ID
180343
Gene Name
60S ribosomal protein L8
Gene Aliases
CELE_B0250.1
Chromosome Location
Chr.V: 20479689 - 20483276 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075539.8 NP_507940.1 1 - 2 290 149

View Full Product Details