Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fipr-27
Entrez Gene ID
4927054
Gene Name
FIP (Fungus-Induced Protein) Related
Gene Aliases
CELE_Y51H4A.32
Chromosome Location
Chr.IV: 16630922 - 16631247 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083267.3 NP_001076736.1 1 - 2 72 135

View Full Product Details