Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EDN3
Entrez Gene ID
403406
Gene Name
endothelin 3
Gene Aliases
PPET3
Chromosome Location
Chr.24: 44024553 - 44054727 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002942.1 NP_001002942.1 1 - 2 123 93
XM_005634662.3 XP_005634719.1 1 - 2 239 93
XM_022408799.1 XP_022264507.1 1 - 2 238 93
GenBank mRNA AB115086.1 - 1 - 2 123 93

View Full Product Details