Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EDN3
Entrez Gene ID
403406
Gene Name
endothelin 3
Gene Aliases
PPET3
Chromosome Location
Chr.24: 44024553 - 44054727 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002942.1 NP_001002942.1 2 - 3 412 84
XM_005634662.3 XP_005634719.1 2 - 3 528 84
XM_022408799.1 XP_022264507.1 2 - 3 527 84
XM_022408800.1 XP_022264508.1 3 - 4 566 84
XM_022408801.1 XP_022264509.1 3 - 4 485 84
GenBank mRNA AB115086.1 - 2 - 3 412 84

View Full Product Details