Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCL28
Entrez Gene ID
448795
Gene Name
C-C motif chemokine ligand 28
Gene Aliases
-
Chromosome Location
Chr.4: 66567139 - 66597310 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005257.1 NP_001005257.1 1 - 2 67 67
GenBank mRNA AB164619.1 - 1 - 2 67 67

View Full Product Details