Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DLA-DQB1
Entrez Gene ID
474862
Gene Name
major histocompatibility complex, class II, DQ beta 1
Gene Aliases
DLA-DQB
Chromosome Location
Chr.12: 2305307 - 2307711 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014381.1 NP_001014403.1 2 - 3 379 119
GenBank mRNA AF043908.1 - 2 - 3 379 119
M90802.1 - 1 - 2 244 119
M90803.1 - 1 - 2 244 119
M90804.1 - 1 - 2 244 119
M90805.1 - 1 - 2 119
S53615.1 - 1 - 2 244 119
Y07945.1 - - 244 119
Y07946.1 - - 244 119
Y07947.1 - - 244 119

Gene Symbol
LOC100856137
Entrez Gene ID
100856137
Gene Name
HLA class II histocompatibility antigen, DQ beta 2 chain
Gene Aliases
DLA-DQB1
Chromosome Location
Chr.12: 2244820 - 2250822 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003639415.4 XP_003639463.1 2 - 3 468 119

View Full Product Details