Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD40LG
Entrez Gene ID
403468
Gene Name
CD40 ligand
Gene Aliases
CD154, TNFSF5, TNLG8B
Chromosome Location
Chr.X: 107001421 - 107023007 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002981.1 NP_001002981.1 4 - 5 689 78
XM_014111303.2 XP_013966778.1 6 - 7 632 78
XM_022415465.1 XP_022271173.1 5 - 6 473 78
XM_022415466.1 XP_022271174.1 6 - 7 676 78
GenBank mRNA AF086711.1 - 4 - 5 410 78
AY333790.1 - 4 - 5 689 78

View Full Product Details