Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SGSH
Entrez Gene ID
403707
Gene Name
N-sulfoglucosamine sulfohydrolase
Gene Aliases
HSS
Chromosome Location
Chr.9: 1540933 - 1547029 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003114.1 NP_001003114.1 - 779 101
GenBank mRNA AF217203.1 - - 779 101

View Full Product Details