The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166047.2 | NP_001159519.2 | 7 - 8 | 869 | 94 |
XM_005636672.3 | XP_005636729.1 | 8 - 9 | 970 | 94 | |
XM_005636673.3 | XP_005636730.1 | 7 - 8 | 912 | 94 | |
XM_022410841.1 | XP_022266549.1 | 7 - 8 | 1157 | 94 | |
GenBank mRNA | AF403244.1 | - | 2 - 3 | 213 | 94 |
GQ497899.1 | - | 7 - 8 | 869 | 94 | |
LN833579.1 | - | 7 - 8 | 839 | 94 |