The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003378.2 | NP_001003378.1 | 1 - 2 | 89 | 114 |
GenBank mRNA | AF494190.1 | - | 1 - 2 | 92 | 114 |