Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLUL
Entrez Gene ID
403443
Gene Name
glutamate-ammonia ligase
Gene Aliases
GS
Chromosome Location
Chr.7: 15730898 - 15740241 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002965.2 NP_001002965.2 - 226 55
GenBank mRNA AF544242.1 - - 230 55
BM537412.1 - - 44 55
DN428718.1 - - 213 55

View Full Product Details