Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CE9
Entrez Gene ID
403794
Gene Name
CE9 protein
Gene Aliases
-
Chromosome Location
Chr.7: 15791793 - 15792956 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003167.1 NP_001003167.1 - 1084 77
GenBank mRNA AJ238951.1 - - 1084 77

View Full Product Details