Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPL23
Entrez Gene ID
403683
Gene Name
ribosomal protein L23
Gene Aliases
RPL17/L23
Chromosome Location
Chr.9: 23397143 - 23403060 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003100.2 NP_001003100.2 1 - 1 215 79
GenBank mRNA AJ388521.1 - 1 - 1 106 79

View Full Product Details