Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFDP1
Entrez Gene ID
403675
Gene Name
craniofacial development protein 1
Gene Aliases
P97
Chromosome Location
Chr.5: 75313165 - 75442686 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287118.1 NP_001274047.1 1 - 2 190 90
XM_022418006.1 XP_022273714.1 1 - 2 217 90
GenBank mRNA AJ388531.1 - 1 - 2 188 90
CV998536.1 - 1 - 2 181 90

View Full Product Details