Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF252
Entrez Gene ID
403421
Gene Name
zinc finger protein 252
Gene Aliases
BC3, ZNF347
Chromosome Location
Chr.13: 38193793 - 38204926 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002954.1 NP_001002954.1 1 - 2 73 64
GenBank mRNA AJ388557.1 - 1 - 2 73 64

View Full Product Details