Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SFRP2
Entrez Gene ID
475471
Gene Name
secreted frizzled related protein 2
Gene Aliases
-
Chromosome Location
Chr.15: 51541197 - 51548765 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002987.2 NP_001002987.1 2 - 3 596 143
GenBank mRNA AJ560716.1 - 2 - 3 596 143

View Full Product Details