Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HTR7
Entrez Gene ID
477762
Gene Name
5-hydroxytryptamine receptor 7
Gene Aliases
-
Chromosome Location
Chr.28: 5438872 - 5524385 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005637529.3 XP_005637586.1 1 - 2 733 72
XM_534958.6 XP_534958.2 1 - 2 730 72
GenBank mRNA AJ628248.1 - 1 - 2 495 72
AJ628249.1 - 1 - 2 495 72

View Full Product Details