Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCNB3
Entrez Gene ID
449494
Gene Name
cyclin B3
Gene Aliases
-
Chromosome Location
Chr.X: 42951030 - 43041616 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005763.1 NP_001005763.1 8 - 9 3658 126
XM_022415500.1 XP_022271208.1 8 - 9 3652 126
XM_022415501.1 XP_022271209.1 8 - 9 3652 126
XM_022415502.1 XP_022271210.1 8 - 9 3652 126
XM_022415503.1 XP_022271211.1 8 - 9 3689 126
GenBank mRNA AJ833648.1 - 8 - 9 3658 126
AM261211.1 - 6 - 7 643 126

View Full Product Details