Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TACR2
Entrez Gene ID
489020
Gene Name
tachykinin receptor 2
Gene Aliases
NK-2R, SKR
Chromosome Location
Chr.4: 20518797 - 20531118 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012617.1 NP_001012635.1 3 - 4 745 77
GenBank mRNA AJ884916.1 - 3 - 4 745 77

View Full Product Details