Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLA2G15
Entrez Gene ID
403403
Gene Name
phospholipase A2 group XV
Gene Aliases
ACS, LLPL, LPLA2, LYPLA3
Chromosome Location
Chr.5: 81269914 - 81280700 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002940.1 NP_001002940.1 2 - 3 308 66
XM_005620606.3 XP_005620663.1 2 - 3 1536 66
GenBank mRNA AY217754.1 - 2 - 3 308 66

View Full Product Details