Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRTDAP
Entrez Gene ID
554216
Gene Name
keratinocyte differentiation associated protein
Gene Aliases
KDAP, KIPV467
Chromosome Location
Chr.1: 117167456 - 117170647 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024642.1 NP_001019813.1 5 - 6 287 97
GenBank mRNA AY999063.1 - 5 - 6 287 97

View Full Product Details