Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FXYD1
Entrez Gene ID
476487
Gene Name
FXYD domain containing ion transport regulator 1
Gene Aliases
PLM
Chromosome Location
Chr.1: 117436239 - 117440619 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195142.1 NP_001182071.1 5 - 6 326 95
NM_001195143.1 NP_001182072.1 5 - 6 326 95
XM_005615754.3 XP_005615811.2 5 - 6 379 95
XM_005615755.3 XP_005615812.2 5 - 6 435 170
XM_014114177.2 XP_013969652.2 5 - 6 620 170
XM_014114183.2 XP_013969658.1 5 - 6 530 170
XM_014114188.2 XP_013969663.1 5 - 6 481 170
GenBank mRNA M63934.1 - 5 - 6 303 95

View Full Product Details