Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACTR1A
Entrez Gene ID
403791
Gene Name
actin related protein 1A
Gene Aliases
-
Chromosome Location
Chr.28: 14972512 - 14990434 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003164.1 NP_001003164.1 6 - 7 742 105
GenBank mRNA S45367.1 - 6 - 7 742 105

View Full Product Details