Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VCAM1
Entrez Gene ID
403982
Gene Name
vascular cell adhesion molecule 1
Gene Aliases
-
Chromosome Location
Chr.6: 49341669 - 49359841 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003298.1 NP_001003298.1 5 - 6 1296 102
XM_005621302.3 XP_005621359.2 5 - 6 1365 102
XM_005621304.2 XP_005621361.1 5 - 6 1217 102
XM_005621305.3 XP_005621362.1 5 - 6 1159 102
GenBank mRNA U32086.1 - 5 - 6 1296 102

View Full Product Details