Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMED11
Entrez Gene ID
403969
Gene Name
glycoprotein 25L
Gene Aliases
GP25L
Chromosome Location
Chr.3: 91435697 - 91452352 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003286.1 NP_001003286.1 3 - 4 356 77
GenBank mRNA X53592.1 - - 356 77

View Full Product Details