Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
APOH
Entrez Gene ID
403945
Gene Name
apolipoprotein H
Gene Aliases
-
Chromosome Location
Chr.9: 14082648 - 14095186 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003265.2 NP_001003265.1 - 430 119
GenBank mRNA X72933.1 - - 430 119

View Full Product Details