Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NR4A1
Entrez Gene ID
403897
Gene Name
nuclear receptor subfamily 4 group A member 1
Gene Aliases
NGFI-B
Chromosome Location
Chr.27: 2947729 - 2965045 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003227.1 NP_001003227.1 1 - 2 63 80
GenBank mRNA X97226.1 - - 63 80

View Full Product Details