Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP3A26
Entrez Gene ID
415127
Gene Name
cytochrome P450 3A26
Gene Aliases
-
Chromosome Location
Chr.6: 9777663 - 9815345 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003338.1 NP_001003338.1 - 1830 82
GenBank mRNA AF547269.1 - - 1830 82

View Full Product Details