Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDK2AP2
Entrez Gene ID
610458
Gene Name
cyclin dependent kinase 2 associated protein 2
Gene Aliases
-
Chromosome Location
Chr.18: 49969630 - 49971781 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_846714.5 XP_851807.1 1 - 2 634 143

View Full Product Details