Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYOZ1
Entrez Gene ID
479250
Gene Name
myozenin 1
Gene Aliases
-
Chromosome Location
Chr.4: 24158532 - 24165416 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001048095.1 NP_001041560.1 1 - 2 74 76
XM_005618876.2 XP_005618933.1 2 - 3 194 76
GenBank mRNA DQ143043.1 - 1 - 2 74 76

View Full Product Details