Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ORMDL3
Entrez Gene ID
480530
Gene Name
ORMDL sphingolipid biosynthesis regulator 3
Gene Aliases
-
Chromosome Location
Chr.9: 22594425 - 22601108 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005624545.3 XP_005624602.1 - 591 110
XM_005624546.3 XP_005624603.1 - 490 110
XM_005624547.3 XP_005624604.1 - 586 110
XM_846217.5 XP_851310.1 - 509 110

View Full Product Details