Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC12A
Entrez Gene ID
477697
Gene Name
C-type lectin domain family 12 member A
Gene Aliases
-
Chromosome Location
Chr.27: 36040574 - 36051249 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_534891.6 XP_534891.2 1 - 2 290 69

View Full Product Details