Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAGLN3
Entrez Gene ID
478562
Gene Name
transgelin 3
Gene Aliases
-
Chromosome Location
Chr.33: 16638371 - 16652418 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_535738.7 XP_535738.1 1 - 2 375 72

View Full Product Details