Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STMN1
Entrez Gene ID
478175
Gene Name
stathmin 1
Gene Aliases
-
Chromosome Location
Chr.2: 74035462 - 74043352 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_535349.6 XP_535349.1 2 - 3 220 68
XM_861033.5 XP_866126.1 2 - 3 137 68

View Full Product Details