Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD46
Entrez Gene ID
475050
Gene Name
ankyrin repeat domain 46
Gene Aliases
-
Chromosome Location
Chr.13: 2384007 - 2415187 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022426585.1 XP_022282293.1 - 641 88
XM_532285.5 XP_532285.2 - 546 88

View Full Product Details