Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPAM1
Entrez Gene ID
475211
Gene Name
sperm adhesion molecule 1
Gene Aliases
-
Chromosome Location
Chr.14: 11704766 - 11723187 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_532443.6 XP_532443.1 1 - 2 136 139

View Full Product Details