Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AREG
Entrez Gene ID
475176
Gene Name
amphiregulin
Gene Aliases
AREGB
Chromosome Location
Chr.13: 62925761 - 62936753 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005628295.3 XP_005628352.1 3 - 4 480 117
XM_022427107.1 XP_022282815.1 3 - 4 531 117
XM_022427108.1 XP_022282816.1 3 - 4 531 117
XM_532410.5 XP_532410.3 3 - 4 604 117

View Full Product Details