Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM17
Entrez Gene ID
474612
Gene Name
transmembrane protein 17
Gene Aliases
-
Chromosome Location
Chr.10: 62355849 - 62361445 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_531842.6 XP_531842.1 3 - 4 401 89

View Full Product Details