Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DOK6
Entrez Gene ID
476173
Gene Name
docking protein 6
Gene Aliases
-
Chromosome Location
Chr.1: 8446332 - 8707600 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022404583.1 XP_022260291.1 - 699 81

View Full Product Details